Assay Design



Gene Transcripts

Gene Symbol
KCNMB2
Entrez Gene ID
100620053
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 2
Gene Aliases
-
Chromosome Location
Chr.1: 226724496 - 226769077 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958237.1 XP_027814038.1 1 - 2 114 72

View Full Product Details