Assay Design



Gene Transcripts

Gene Symbol
SS18L1
Entrez Gene ID
101113515
Gene Name
SS18L1 subunit of BAF chromatin remodeling complex
Gene Aliases
-
Chromosome Location
Chr.13: 57376707 - 57404471 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976979.1 XP_027832780.1 8 - 9 991 54

View Full Product Details