Assay Design



Gene Transcripts

Gene Symbol
NRBF2
Entrez Gene ID
101122660
Gene Name
nuclear receptor binding factor 2
Gene Aliases
-
Chromosome Location
Chr.25: 19111745 - 19140563 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021396.3 XP_004021445.1 2 - 3 311 86

View Full Product Details