Assay Design



Gene Transcripts

Gene Symbol
ANKRD22
Entrez Gene ID
101120533
Gene Name
ankyrin repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.22: 12221505 - 12253552 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020017.4 XP_004020066.1 3 - 4 524 65

View Full Product Details