Assay Design



Gene Transcripts

Gene Symbol
PEX12
Entrez Gene ID
101116741
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
-
Chromosome Location
Chr.11: 48998848 - 49003007 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012458.4 XP_004012507.1 1 - 2 807 94

View Full Product Details