Assay Design



Gene Transcripts

Gene Symbol
SYNPO2
Entrez Gene ID
101110330
Gene Name
synaptopodin 2
Gene Aliases
-
Chromosome Location
Chr.6: 7771962 - 7977627 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009590.4 XP_004009639.2 3 - 4 1173 80
XM_012179415.3 XP_012034805.2 3 - 4 1173 80
XM_012179416.3 XP_012034806.2 3 - 4 1171 80

View Full Product Details