Assay Design



Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
101107539
Gene Name
ASPSCR1 tether for SLC2A4, UBX domain containing
Gene Aliases
-
Chromosome Location
Chr.11: 12189893 - 12218236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098906.2 XP_014954392.2 15 - 16 1697 119
XM_027974287.1 XP_027830088.1 15 - 16 1905 119
XM_027974289.1 XP_027830090.1 14 - 15 1620 119

View Full Product Details