Assay Design



Gene Transcripts

Gene Symbol
KCNC1
Entrez Gene ID
101113880
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
-
Chromosome Location
Chr.15: 37641795 - 37686184 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016119.4 XP_004016168.1 3 - 4 2305 96

View Full Product Details