Assay Design



Gene Transcripts

Gene Symbol
TXNDC17
Entrez Gene ID
100302359
Gene Name
thioredoxin domain containing 17
Gene Aliases
-
Chromosome Location
Chr.11: 38120361 - 38122812 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163057.1 NP_001156529.1 3 - 4 335 82
GenBank mRNA FJ969407.1 - 3 - 4 335 82

View Full Product Details