Assay Design



Gene Transcripts

Gene Symbol
FGF22
Entrez Gene ID
101114608
Gene Name
fibroblast growth factor 22
Gene Aliases
-
Chromosome Location
Chr.5: 45369989 - 45373371 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008762.4 XP_004008811.3 1 - 2 217 70

View Full Product Details