Assay Design



Gene Transcripts

Gene Symbol
RBM38
Entrez Gene ID
101104417
Gene Name
RNA binding motif protein 38
Gene Aliases
-
Chromosome Location
Chr.13: 61083182 - 61098770 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023341.4 XP_004023390.4 1 - 2 407 79

View Full Product Details