Assay Design



Gene Transcripts

Gene Symbol
NOVA2
Entrez Gene ID
101120187
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
-
Chromosome Location
Chr.14: 56936899 - 56963221 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978509.1 XP_027834310.1 3 - 4 408 66

View Full Product Details