Assay Design



Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
101116077
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.13: 56171505 - 56176053 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976932.1 XP_027832733.1 3 - 4 504 80

View Full Product Details