Assay Design



Gene Transcripts

Gene Symbol
ARPC5
Entrez Gene ID
101106701
Gene Name
actin related protein 2/3 complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.12: 69246271 - 69256570 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013863.3 XP_004013912.2 3 - 4 596 89

View Full Product Details