Assay Design



Gene Transcripts

Gene Symbol
NKX6-1
Entrez Gene ID
554330
Gene Name
NK6 homeobox 1
Gene Aliases
NKX6.1
Chromosome Location
Chr.6: 109590908 - 109595927 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971409.1 XP_027827210.1 2 - 3 840 86
GenBank mRNA DQ054803.1 - 1 - 2 168 86

View Full Product Details