Assay Design



Gene Transcripts

Gene Symbol
CSN1S2
Entrez Gene ID
443383
Gene Name
alpha-S2-casein
Gene Aliases
-
Chromosome Location
Chr.6: 94793750 - 94810274 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009363.1 NP_001009363.1 13 - 14 525 98
GenBank mRNA GU169085.1 - 13 - 14 503 98
GU169086.1 - 13 - 14 503 98
GU169087.1 - 13 - 14 503 98
GU169088.1 - 13 - 14 503 98
X03238.1 - - 525 98

View Full Product Details