Assay Design



Gene Transcripts

Gene Symbol
MSTN
Entrez Gene ID
443449
Gene Name
myostatin
Gene Aliases
GDF-8, GDF8
Chromosome Location
Chr.2: 129059547 - 129066303 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009428.3 NP_001009428.1 1 - 2 580 66
GenBank mRNA AF019622.1 - 1 - 2 372 66
AM992883.1 - 1 - 2 481 66
AM992884.1 - 1 - 2 481 66
FM207636.1 - 1 - 2 482 66
KM371731.1 - 1 - 2 580 66

View Full Product Details