Assay Design



Gene Transcripts

Gene Symbol
RHOB
Entrez Gene ID
100158235
Gene Name
ras homolog family member B
Gene Aliases
-
Chromosome Location
Chr.3: 29654512 - 29656639 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127673.3 NP_001121145.1 1 - 1 560 79
GenBank mRNA EU664600.1 - 1 - 1 517 79

View Full Product Details