Assay Design



Gene Transcripts

Gene Symbol
HSD11B1
Entrez Gene ID
443409
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.12: 77036851 - 77105871 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009395.2 NP_001009395.1 4 - 5 581 84
XM_012186882.2 XP_012042272.1 5 - 6 804 84
XM_027975568.1 XP_027831369.1 7 - 8 1084 84
XM_027975569.1 XP_027831370.1 6 - 7 1444 84
XM_027975570.1 XP_027831371.1 6 - 7 913 84
GenBank mRNA X69561.1 - - 584 84

View Full Product Details