Assay Design



Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
100145884
Gene Name
vasoactive intestinal peptide
Gene Aliases
-
Chromosome Location
Chr.8: 83836510 - 83845502 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126368.1 NP_001119840.1 3 - 4 331 73
XM_012182368.3 XP_012037758.1 4 - 5 578 73
GenBank mRNA EU016225.1 - 3 - 4 331 73

View Full Product Details