Assay Design



Gene Transcripts

Gene Symbol
NPVF
Entrez Gene ID
100127217
Gene Name
neuropeptide VF precursor
Gene Aliases
RFRP
Chromosome Location
Chr.4: 77511001 - 77514323 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127268.1 NP_001120740.1 1 - 2 136 194
GenBank mRNA EF494241.1 - 1 - 2 136 194

View Full Product Details