Assay Design



Gene Transcripts

Gene Symbol
MYOG
Entrez Gene ID
443158
Gene Name
myogenin
Gene Aliases
-
Chromosome Location
Chr.12: 253164 - 255189 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174109.1 NP_001167580.1 1 - 2 474 96
GenBank mRNA GU550517.1 - 1 - 2 474 96

View Full Product Details