Assay Design



Gene Transcripts

Gene Symbol
SLAMF9
Entrez Gene ID
101123601
Gene Name
SLAM family member 9
Gene Aliases
-
Chromosome Location
Chr.1: 117815912 - 117818860 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302661.2 NP_001289590.2 2 - 3 387 60

View Full Product Details