Assay Design



Gene Transcripts

Gene Symbol
SPA17
Entrez Gene ID
443067
Gene Name
sperm autoantigenic protein 17
Gene Aliases
SP17
Chromosome Location
Chr.21: 29152895 - 29165213 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009738.1 NP_001009738.1 4 - 5 396 103
XM_012101537.3 XP_011956927.1 4 - 5 103
GenBank mRNA AF179926.1 - 4 - 5 396 103

View Full Product Details