Assay Design



Gene Transcripts

Gene Symbol
FCGR3A
Entrez Gene ID
100192433
Gene Name
Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
Gene Aliases
-
Chromosome Location
Chr.1: 120125218 - 120136004 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139454.1 NP_001132926.1 5 - 6 770 72
GenBank mRNA EU589390.1 - 5 - 6 770 72

View Full Product Details