Assay Design



Gene Transcripts

Gene Symbol
CCND2
Entrez Gene ID
100147799
Gene Name
cyclin D2
Gene Aliases
-
Chromosome Location
Chr.3: 226563141 - 226585958 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127290.1 NP_001120762.1 3 - 4 574 67
GenBank mRNA EU626221.1 - 3 - 4 574 67

View Full Product Details