Assay Design



Gene Transcripts

Gene Symbol
WNT2
Entrez Gene ID
100126577
Gene Name
Wnt family member 2
Gene Aliases
-
Chromosome Location
Chr.4: 57434623 - 57480646 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195319.1 NP_001182248.1 4 - 5 1150 92

View Full Product Details