Assay Design



Gene Transcripts

Gene Symbol
NDUFB6
Entrez Gene ID
100462693
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
-
Chromosome Location
Chr.2: 109909296 - 109923951 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185123.1 NP_001172052.1 1 - 3 181 86
GenBank mRNA HM017955.1 - 1 - 3 181 86

View Full Product Details