Assay Design



Gene Transcripts

Gene Symbol
SCGB3A2
Entrez Gene ID
780461
Gene Name
secretoglobin family 3A member 2
Gene Aliases
-
Chromosome Location
Chr.5: 61616471 - 61619948 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195315.1 NP_001182244.1 2 - 3 342 79
GenBank mRNA DQ223549.1 - 2 - 3 342 79

View Full Product Details