Assay Design



Gene Transcripts

Gene Symbol
NPAS4
Entrez Gene ID
101113970
Gene Name
neuronal PAS domain protein 4
Gene Aliases
-
Chromosome Location
Chr.21: 46243994 - 46248721 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287463.1 NP_001274392.1 4 - 5 704 65
GenBank mRNA HF912412.1 - 4 - 5 704 65

View Full Product Details