Assay Design



Gene Transcripts

Gene Symbol
PI3
Entrez Gene ID
641305
Gene Name
peptidase inhibitor 3
Gene Aliases
TOM
Chromosome Location
Chr.13: 76612295 - 76613869 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035224.1 NP_001030301.1 1 - 2 84 91

View Full Product Details