Assay Design



Gene Transcripts

Gene Symbol
CIB1
Entrez Gene ID
100141300
Gene Name
calcium and integrin binding 1
Gene Aliases
Calmyrin
Chromosome Location
Chr.18: 19039357 - 19042704 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114765.1 NP_001108237.1 5 - 6 517 93
GenBank mRNA EU430744.1 - 5 - 6 517 93

View Full Product Details