Assay Design



Gene Transcripts

Gene Symbol
PRP2
Entrez Gene ID
100127213
Gene Name
prolactin-related protein-2
Gene Aliases
-
Chromosome Location
Chr.20: 37219130 - 37231859 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112819.1 NP_001106290.1 2 - 3 288 93
GenBank mRNA AB231298.1 - 2 - 3 288 93

View Full Product Details