Assay Design



Gene Transcripts

Gene Symbol
RRAGC
Entrez Gene ID
100302070
Gene Name
Ras related GTP binding C
Gene Aliases
-
Chromosome Location
Chr.1: 14126398 - 14139367 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161867.1 NP_001155339.1 5 - 6 903 107
GenBank mRNA FJ943975.1 - 5 - 6 903 107

View Full Product Details