Assay Design



Gene Transcripts

Gene Symbol
ST6GAL1
Entrez Gene ID
100913159
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.1: 218708272 - 218847806 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267880.1 NP_001254809.1 4 - 5 956 103
XM_027964968.1 XP_027820769.1 7 - 8 1613 103
XM_027965018.1 XP_027820819.1 6 - 7 1464 103
XM_027965050.1 XP_027820851.1 5 - 6 1233 103
XM_027965081.1 XP_027820882.1 5 - 6 6539 103
GenBank mRNA JQ423938.1 - 4 - 5 956 103

Gene Symbol
LOC114113140
Entrez Gene ID
114113140
Gene Name
beta-galactoside alpha-2,6-sialyltransferase 1-like
Gene Aliases
-
Chromosome Location
Chr.2: 201226912 - 201229333 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964830.1 XP_027820631.1 2 - 2 342 103

View Full Product Details