Assay Design



Gene Transcripts

Gene Symbol
SLC2A3
Entrez Gene ID
443308
Gene Name
solute carrier family 2 member 3
Gene Aliases
GLUT-3
Chromosome Location
Chr.3: 222584545 - 222597190 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009770.1 NP_001009770.1 9 - 10 1388 81
GenBank mRNA L39214.1 - 9 - 10 1388 81

View Full Product Details