Assay Design



Gene Transcripts

Gene Symbol
SRI
Entrez Gene ID
100302556
Gene Name
sorcin
Gene Aliases
-
Chromosome Location
Chr.4: 79317185 - 79329637 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163603.1 NP_001157075.1 6 - 7 544 110
GenBank mRNA FJ937948.1 - 6 - 7 544 110

View Full Product Details