Assay Design



Gene Transcripts

Gene Symbol
IL21
Entrez Gene ID
100862657
Gene Name
interleukin 21
Gene Aliases
-
Chromosome Location
Chr.17: 41070843 - 41078033 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256817.1 NP_001243746.1 2 - 3 208 105
GenBank mRNA HE652877.1 - 2 - 3 208 105

View Full Product Details