Assay Design



Gene Transcripts

Gene Symbol
LHB
Entrez Gene ID
443395
Gene Name
luteinizing hormone beta polypeptide
Gene Aliases
-
Chromosome Location
Chr.14: 59018344 - 59019413 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009380.1 NP_001009380.1 2 - 2 121 75
GenBank mRNA X52488.1 - - 121 75

View Full Product Details