Assay Design



Gene Transcripts

Gene Symbol
CCR10
Entrez Gene ID
678671
Gene Name
C-C motif chemokine receptor 10
Gene Aliases
-
Chromosome Location
Chr.11: 20396190 - 20398153 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040287.1 NP_001035377.1 2 - 2 125 102
GenBank mRNA DQ133532.1 - 2 - 2 125 102

View Full Product Details