Assay Design



Gene Transcripts

Gene Symbol
RELA
Entrez Gene ID
101102191
Gene Name
RELA proto-oncogene, NF-kB subunit
Gene Aliases
-
Chromosome Location
Chr.21: 45624264 - 45634524 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959295.1 XP_027815096.1 9 - 10 2665 107

View Full Product Details