Assay Design



Gene Transcripts

Gene Symbol
CFAP300
Entrez Gene ID
101121882
Gene Name
cilia and flagella associated protein 300
Gene Aliases
C15H11orf70
Chromosome Location
Chr.15: 7036010 - 7064710 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979145.1 XP_027834946.1 6 - 7 700 105

View Full Product Details