Assay Design



Gene Transcripts

Gene Symbol
PSENEN
Entrez Gene ID
101120856
Gene Name
presenilin enhancer, gamma-secretase subunit
Gene Aliases
-
Chromosome Location
Chr.14: 49187570 - 49188974 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015186.3 XP_004015235.1 2 - 3 294 81
XM_004015187.4 XP_004015236.1 2 - 3 206 81

View Full Product Details