Assay Design



Gene Transcripts

Gene Symbol
PLA2G2F
Entrez Gene ID
101106664
Gene Name
phospholipase A2 group IIF
Gene Aliases
-
Chromosome Location
Chr.2: 262051333 - 262062418 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093732.2 XP_014949218.2 2 - 3 267 98

View Full Product Details