Assay Design



Gene Transcripts

Gene Symbol
PRSS50
Entrez Gene ID
101103019
Gene Name
serine protease 50
Gene Aliases
-
Chromosome Location
Chr.19: 54730903 - 54736309 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957902.1 XP_027813703.1 6 - 7 1332 71

View Full Product Details