Assay Design



Gene Transcripts

Gene Symbol
PEF1
Entrez Gene ID
101118674
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 251453543 - 251471319 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005038.3 XP_004005087.2 2 - 3 418 69

View Full Product Details