Assay Design



Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
101107276
Gene Name
ependymin related 1
Gene Aliases
-
Chromosome Location
Chr.4: 55938274 - 55973486 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968603.1 XP_027824404.1 2 - 3 810 96

View Full Product Details