Assay Design



Gene Transcripts

Gene Symbol
SETBP1
Entrez Gene ID
101108271
Gene Name
SET binding protein 1
Gene Aliases
-
Chromosome Location
Chr.23: 49685992 - 50094479 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960948.1 XP_027816749.1 4 - 5 4211 85
XM_027960949.1 XP_027816750.1 4 - 5 4245 85

View Full Product Details