Assay Design



Gene Transcripts

Gene Symbol
HTR7
Entrez Gene ID
101122994
Gene Name
5-hydroxytryptamine receptor 7
Gene Aliases
-
Chromosome Location
Chr.22: 14366748 - 14464671 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020028.4 XP_004020077.2 1 - 2 539 67

View Full Product Details