Assay Design



Gene Transcripts

Gene Symbol
HAO2
Entrez Gene ID
101110803
Gene Name
hydroxyacid oxidase 2
Gene Aliases
-
Chromosome Location
Chr.1: 101895564 - 101925651 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002387.4 XP_004002436.2 4 - 5 862 123
XM_012179575.3 XP_012034965.2 4 - 5 839 123
XM_012179579.3 XP_012034969.2 5 - 6 945 123

View Full Product Details